Skip to Content
Merck
All Photos(1)

Key Documents

70620

Sigma-Aldrich

Ddz-Cys(StBu)-OH (dicyclohexylammonium) salt

purum, ≥98.0% (HPLC)

Synonym(s):

Ddz-S-tert-butylthio-L-cysteine (dicyclohexylammonium) salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H29NO6S2 · C12H23N
CAS Number:
Molecular Weight:
612.88
MDL number:
UNSPSC Code:
12352200

grade

purum

Assay

≥98.0% (HPLC)

SMILES string

C1CCC(CC1)NC2CCCCC2.COc3cc(OC)cc(c3)C(C)(C)OC(=O)N[C@@H](CSSC(C)(C)C)C(O)=O


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service