Skip to Content
Merck
All Photos(1)

Documents

69534

Supelco

Derivatizing agents, Set for GC: Alcohols with boron trifluoride

for GC derivatization, LiChropur

Synonym(s):

Alcohols with boron trifluoride, derivatizing agents, set for GC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
NA.22

grade

for GC derivatization

form

liquid

quality

LiChropur

storage temp.

2-8°C

Components

set contains:
05576 boron trifluoride [~10% (~1.3 M) in ethanol]
15718 boron trifluoride [~14% (~2 M) in 1-propanol]
15716 boron trifluoride [~10% (~1.3 M) in methanol]
83253 boron trifluoride [~10% (~1.3 M) in butanol]
10 mL each

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT RE 2 Inhalation - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system, Eyes,Central nervous system, Kidney, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service