Skip to Content
Merck
All Photos(1)

Key Documents

68853

Supelco

Isoquinoline

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7N
CAS Number:
Molecular Weight:
129.16
Beilstein:
107549
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.623 (lit.)

bp

242-243 °C (lit.)

mp

26-28 °C (lit.)

density

1.099 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-8°C

SMILES string

c1ccc2cnccc2c1

InChI

1S/C9H7N/c1-2-4-9-7-10-6-5-8(9)3-1/h1-7H

InChI key

AWJUIBRHMBBTKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric synthesis of isoquinoline alkaloids.
Maria Chrzanowska et al.
Chemical reviews, 104(7), 3341-3370 (2004-07-15)
Beta-phenylethylamines and the isoquinoline alkaloids.
K W Bentley
Natural product reports, 18(2), 148-170 (2001-05-05)
Kakali Bhadra et al.
Mini reviews in medicinal chemistry, 10(13), 1235-1247 (2010-08-19)
Alkaloids are a group of natural products with unmatched chemical diversity and biological relevance forming potential quality pools in drug screening. The molecular aspects of their interaction with many cellular macromolecules like DNA, RNA and proteins are being currently investigated
Fumihiko Sato et al.
Proceedings of the Japan Academy. Series B, Physical and biological sciences, 89(5), 165-182 (2013-05-15)
Plants produce a variety of secondary metabolites that possess strong physiological activities. Unfortunately, however, their production can suffer from a variety of serious problems, including low levels of productivity and heterogeneous quality, as well as difficulty in raw material supply.
Yusuke Ohta
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 130(7), 925-936 (2010-07-08)
A copper-catalyzed synthesis of 2-(aminomethyl)indole through domino three-component coupling-cyclization has been developed. This reaction proceeds through Mannich-type coupling of 2-ethynylanilines, aldehydes, and secondary amines, followed by hydroamination. This indole formation was applicable to the synthesis of 4-methylene-2,3,4,9-tetrahyro-1H-pyrido[3,4-b]indoles and 5,6,7,8-tetrahydrobenzo[e]indolo[2,3-c]azepines via

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service