Skip to Content
Merck
All Photos(1)

Key Documents

68500

Supelco

Cinnamyl isobutyrate

analytical standard

Synonym(s):

Isobutyric acid cinnamyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCO2CH2CH=CHC6H5
CAS Number:
Molecular Weight:
204.26
EC Number:
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥96.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.523-1.528
n20/D 1.524 (lit.)

bp

254 °C (lit.)

density

1.008 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CC(C)C(=O)OC\C=C\c1ccccc1

InChI

1S/C13H16O2/c1-11(2)13(14)15-10-6-9-12-7-4-3-5-8-12/h3-9,11H,10H2,1-2H3/b9-6+

InChI key

KLKQSZIWHVEARN-RMKNXTFCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cinnamyl isobutyrate (CIB) is an aroma component,[1] naturally occurring in the essential oil of cinnamon bark.[2] This fragrance ingredient is widely found in fragrances used in shampoos, decorative cosmetics, fine fragrances, toilet soaps and other toiletries, non-cosmetic products including household cleaners, detergents, etc.[3]

Application

Cinnamyl isobutyrate may be used as an analytical reference standard for the quantification of the analyte in watermelon (EG) flavored hookah tobacco[1] and leaf oil of Heteropyxis dehniae Suess.[4] using gas chromatography coupled to mass spectrometry (GC-MS).

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cinnamyl isobutyrate decreases plasma glucose levels and total energy intake from a standardized breakfast: A randomized, crossover intervention
Hochkogler MC, et al.
Molecular Nutrition And Food Research, 62(17), 1701038-1701038 (2018)
Composition and bioactivity of the leaf essential oil of Heteropyxis dehniae from Zimbabwe
Sibanda S, et al.
Journal of Ethnopharmacology, 92(1), 107-111 (2004)
Fragrance material review on cinnamyl isobutyrate
Bhatia PS, et al.
Food And Chemical Toxicology, 45(1), S74-S77 (2007)
Specific features of human monocytes activation by monophosphoryl lipid A
Chentouh R, et al.
Scientific reports, 8(1), 1-13 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service