Skip to Content
Merck
All Photos(2)

Key Documents

67652

Millipore

Acinetobacter Millichrome plus Supplement

suitable for microbiology

Synonym(s):

Acinetobacter chromogenic supplement

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171614
NACRES:
NA.74

sterility

sterile; aseptically filled

Quality Level

form

dry powder

shelf life

limited shelf life, expiry date on the label

technique(s)

microbiological culture: suitable

solubility

sterile water: soluble (1 vial is sufficient for 5 L of final media)

application(s)

microbiology
pharmaceutical
water monitoring

shipped in

ambient

storage temp.

2-8°C

suitability

selective for bacteria: Acinetobacter spp

General description

Acinetobacter Millichrome plus Supplement is a selective media supplement for the growth promotion of Acinetobacter. Millichrome chromogenic media is a specialized type of culture medium used to detect and isolate specific groups of bacteria based on their ability to produce enzymes with chromogenic substrates. This Acinetobacter supplement is composed of growth and regulatory factors. One vial of the supplement is sufficient for 5 L of final media.

Application

Acinetobacter Millichrome plus Supplement is a selective chromogenic media supplement intended for use in the qualitative detection, differentiation and presumptive identification of Acinetobacter from various samples.

Components

Composition:
(1 vial is sufficient for 5 L of final media)
Growth and regulator factors 20 mL

Preparation Note

Add 4.0 mL of the liquid Acinetobacter Millichrome plus Supplement into 1 L Acinetobacter Millichrome plus Agar base.

Analysis Note

The Acinetobacter is detected by a characteristic red colour after 24 h at an optimal growth temperature of 37°C, while most other Gram-negative bacteria form blue or inhibited after incubation at the same temperature.

Legal Information

MilliChrome is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Selective media enable faster results and visual confirmation for the detection, identification, and enumeration of microorganisms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service