Skip to Content
Merck
All Photos(1)

Key Documents

67560

Sigma-Aldrich

1-Methylimidazole

puriss., ≥99.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N2
CAS Number:
Molecular Weight:
82.10
Beilstein:
105197
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor pressure

0.4 mmHg ( 20 °C)

grade

puriss.

Assay

≥99.0% (GC)

autoignition temp.

977 °F

expl. lim.

15.7 %

refractive index

n20/D 1.495 (lit.)
n20/D 1.497

bp

198 °C (lit.)

mp

−6 °C (lit.)

density

1.03 g/mL at 25 °C (lit.)

SMILES string

Cn1ccnc1

InChI

1S/C4H6N2/c1-6-3-2-5-4-6/h2-4H,1H3

InChI key

MCTWTZJPVLRJOU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Catalyst for the mild analytical acetylation of hydroxy compounds

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K.A. Connors et al.
Analytical Chemistry, 50, 1542-1542 (1978)
L.A. Dee et al.
Analytical Chemistry, 52, 572-572 (1980)
Pengfei Zhang et al.
Journal of chromatography. A, 1218(22), 3459-3465 (2011-04-19)
The development of mixed-mode stationary phase to achieve multiple separation capabilities in one column is very important for high performance liquid chromatography. In this paper, a new specific stationary phase based on grafting N-methylimidazolium to a monolithic silica column was
Bhaskar Sharma et al.
The journal of physical chemistry. A, 115(10), 1971-1984 (2011-02-22)
Quantum chemical [MP2(FULL)/6-311++G-(d,p)] calculations are done on the binding of hydrated Li(+), Na(+), K(+), Mg(2+), Cu(+), and Zn(2+) metal ions with biologically relevant heteroaromatics such as imidazole and methylimidazole. The computed interaction energies are found to be in good agreement
Zhan-Yong Wang et al.
The Journal of organic chemistry, 77(15), 6608-6614 (2012-07-19)
A well-defined NHC-Pd(II)-Im complex 1 was found to be an effective catalyst for the Suzuki-Miyaura coupling of aryl sulfonates including tosylates and phenylsulfonates with arylboronic acids, giving the desired coupling products in good to high yields. Acceptable yields can also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service