Skip to Content
Merck
All Photos(1)

Key Documents

66964

Millipore

Differential ID Membrane

suitable for microbiology, pinkish colored, sterile, diameter 70 mm

Synonym(s):

Indicator membrane, ID Membranes

Sign Into View Organizational & Contract Pricing

Select a Size

25 DISCS
R$166.00

R$166.00


Estimated to ship onApril 16, 2025


Request more information

Select a Size

Change View
25 DISCS
R$166.00

About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

R$166.00


Estimated to ship onApril 16, 2025


Request more information

membrane size

70 mm

Quality Level

sterility

sterile

shelf life

limited shelf life, expiry date on the label

storage condition

protect from light

technique(s)

microbe id | utilization test: suitable

application(s)

cleaning products
clinical testing
cosmetics
environmental
food and beverages
personal care
pharmaceutical

microbiology

storage temp.

2-8°C

suitability

Citrobacter spp.
Enterobacter spp.
Escherichia coli
Klebsiella spp.
enterobacteriaceae

General description

Differential ID Membrane is a substrate-carrying ID membrane used for rapid identification and differentiation of lactose fermenting and lactose non-fermenting enteric bacteria through characteristic color reactions. The membranes contain chromogenic substrates, which serve as the basis for microbe differentiation by color. The principle is based on a specific substrate enzyme reaction in which specific lactose fermenting organisms will exhibit dark pink color.

Application

Differential ID Membrane is used for rapid detection & confirmation of lactose fermenting and lactose non-fermenting enteric bacteria in water and food samples. They find their application in various sectors such as the food and dairy industry, water industry, pharmaceutical laboratory testing, cosmetic industry, environmental and sanitary testing, clinical diagnostic, etc.

Analysis Note

Escherichia coli (25922)Dark pinkEscherichia coli (8739)Dark pinkStaphylococcus aureus (25923)colorlessSalmonella Typhimurium (14028)colorless

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service