Skip to Content
Merck
All Photos(1)

Key Documents

66070

Supelco

3-Methyl-1-butene

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
R$776.00

R$776.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

Select a Size

Change View
10 ML
R$776.00

About This Item

Linear Formula:
(CH3)2CHCH=CH2
CAS Number:
Molecular Weight:
70.13
Beilstein:
505980
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

R$776.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

2-methyl-1-butene and 1-pentene, residual

refractive index

n20/D 1.364
n20/D 1.366

bp

20 °C (lit.)

density

0.627 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care
petroleum

format

neat

storage temp.

−20°C

SMILES string

CC(C)C=C

InChI

1S/C5H10/c1-4-5(2)3/h4-5H,1H2,2-3H3

InChI key

YHQXBTXEYZIYOV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

10 mL septum vial

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-70.6 °F - closed cup

Flash Point(C)

-57 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service