Skip to Content
Merck
All Photos(3)

Key Documents

65365

Supelco

(S)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride

for chiral derivatization, LiChropur, ≥99.0%

Synonym(s):

(S)-(+)-MTPA-Cl, Mosher’s acid chloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
R$1,419.00
500 MG
R$5,323.00

R$1,419.00


Estimated to ship onApril 12, 2025


Request a Bulk Order

Select a Size

Change View
100 MG
R$1,419.00
500 MG
R$5,323.00

About This Item

Linear Formula:
C6H5C(OCH3)(CF3)COCl
CAS Number:
Molecular Weight:
252.62
Beilstein:
3591562
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

R$1,419.00


Estimated to ship onApril 12, 2025


Request a Bulk Order

grade

for chiral derivatization

Quality Level

Assay

≥99.0% (AT)
≥99.0%

optical activity

[α]20/D +132±2°, c = 6% in chloroform

optical purity

enantiomeric ratio: ≥99.5:0.5 (GC)

quality

LiChropur

technique(s)

HPLC: suitable

refractive index

n20/D 1.469 (lit.)
n20/D 1.469

bp

213-214 °C (lit.)

density

1.35 g/mL at 25 °C (lit.)

shipped in

wet ice

storage temp.

−20°C

SMILES string

CO[C@@](C(Cl)=O)(c1ccccc1)C(F)(F)F

InChI

1S/C10H8ClF3O2/c1-16-9(8(11)15,10(12,13)14)7-5-3-2-4-6-7/h2-6H,1H3/t9-/m1/s1

InChI key

PAORVUMOXXAMPL-SECBINFHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Determination of absolute configuration of stereogenic carbinol carbons; doi:10.1038/nprot.2007.354[1]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K J Miller et al.
Journal of chromatography, 307(2), 335-342 (1984-05-11)
High-performance liquid chromatography (HPLC) was employed for resolution of enantiomers of chiral ring-substituted 1-phenyl-2- aminopropanes (amphetamines) and 1-phenylethylamine following derivatization with four chiral reagents: (R)-(+)-1-phenylethyl isocyanate ( PEIC ), (-)-alpha-methoxy-alpha-(trifluoromethyl)phenylacetyl chloride ( MTPA X Cl), 2,3,4,6-tetra-O-acetyl-beta-D-glucopyranosyl isothiocyanate ( GITC )
Thomas R Hoye et al.
Nature protocols, 2(10), 2451-2458 (2007-10-20)
This protocol details the most commonly used nuclear magnetic resonance (NMR)-based method for deducing the configuration of otherwise unknown stereogenic, secondary carbinol (alcohol) centers (R1R2CHOH (or the analogous amines where OH is replaced by NH2)). This 'Mosher ester analysis' relies

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service