Key Documents
63653
Supelco
Punch for ion-selective membranes
for ion-selective electrodes, Selectophore
About This Item
UNSPSC Code:
26111700
NACRES:
NB.61
grade
for ion-selective electrodes
product line
Selectophore
General description
Visit our Sensor Applications portal to learn more.
containing 4 modules with inside diameters of 3, 5, 7 and 8 mm
Legal Information
Selectophore is a trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)