Skip to Content
Merck
All Photos(2)

Key Documents

62707

Supelco

Bergaptol

analytical standard

Synonym(s):

4-Hydroxy7H-furo[3,2-g][1]benzopyran-7-one, 4-Hydroxybergapten, 5-Hydroxy-6,7-furanocoumarin, 5-Hydroxypsoralen, NSC 341958

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H6O4
CAS Number:
Molecular Weight:
202.16
Beilstein:
190104
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Assay

≥98.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

O=C(C=C1)OC2=C1C(O)=C(C=CO3)C3=C2

InChI

1S/C11H6O4/c12-10-2-1-6-9(15-10)5-8-7(11(6)13)3-4-14-8/h1-5,13H

InChI key

GIJHDGJRTUSBJR-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Salvatore Multari et al.
Food research international (Ottawa, Ont.), 134, 109228-109228 (2020-06-11)
Citrus are fruit crops that are widely cultivated in Southern Italy. The phytochemical profile of citrus depends on the species, the tissue and the ripening stage. This study aimed to perform a comprehensive analysis of the phenolic compounds and carotenoids
Melissa M Melough et al.
Journal of agricultural and food chemistry, 65(24), 5049-5055 (2017-06-06)
Furocoumarins are a class of photoactive compounds found in several plant species and may be responsible for the observed association between consumption of citrus products and the risk of skin cancer. Furocoumarin contents of several foods have been reported previously
Xinjing Dou et al.
Metabolites, 10(3) (2020-03-04)
Food adulteration is a challenge faced by consumers and researchers. Due to DNA fragmentation during oil processing, it is necessary to discover metabolic markers alternative to DNA for adulteration detection of edible oils. However, the contents of metabolic markers vary
Yelena Guttman et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 137, 111135-111135 (2020-01-23)
Furanocoumarins are the main compounds responsible for the food-drug interactions known as the grapefruit effect, which is caused by the inhibition of CYP3A4-mediated drug metabolism. We evaluated the effects of two new, low-furanocoumarin grapefruit cultivars on CYP3A4 activity and the
Nicole Raquet et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 68, 257-266 (2014-04-01)
Furocoumarins (FCs) are natural constituents widely occurring in plants used as food or in phytomedicines, cosmetics, etc. Some FCs exert dermal photo-toxicity and -genotoxicity when combined with UVA irradiation. For a few congeners, skin tumor formation has been described in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service