Skip to Content
Merck
All Photos(1)

Key Documents

61924

Supelco

Bis(4-methyl-2-pentyl)phthalate

analytical standard

Synonym(s):

Bis(1,3-dimethylbutyl)phthalate, Phthalic acid bis(4-methyl-2-pentyl) ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H30O4
CAS Number:
Molecular Weight:
334.45
Beilstein:
3368171
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

application(s)

environmental

format

neat

storage temp.

2-8°C

SMILES string

O=C(OC(CC(C)C)C)C1=CC=CC=C1C(OC(CC(C)C)C)=O

InChI

1S/C20H30O4/c1-13(2)11-15(5)23-19(21)17-9-7-8-10-18(17)20(22)24-16(6)12-14(3)4/h7-10,13-16H,11-12H2,1-6H3

InChI key

UAFXUVUVOTXFND-UHFFFAOYSA-N

General description

Bis(4-methyl-2-pentyl)phthalate (BMPP), a phthalate ester (PAE), is extensively used as a plasticizer to improve the flexibility and durability of the end products.

Application

Bis(4-methyl-2-pentyl)phthalate is used as a reference standard for the determination of the analyte in various samples by analytical methods.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The influence of facility agriculture production on phthalate esters distribution in black soils of northeast China
Zhang Y, et al.
The Science of the Total Environment, 506, 118-125 (2015)
Phthalate esters in soil, plastic film, and vegetable from greenhouse vegetable production bases in Beijing, China: concentrations, sources, and risk assessment
Li C, et al.
The Science of the Total Environment, 568, 1037-1043 (2016)
Ji-Cai Fan et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 37(11), 1897-1905 (2020-09-23)
This study describes the migration of 20 phthalates from breast milk storage bags to breast milk after storage frozen for 1 to 6 months. Analysis of the investigated phthalates was performed by GC-MS/MS. Nine phthalates were found in some of the
Solid-phase extraction based on chloromethylated polystyrene magnetic nanospheres followed by gas chromatography with mass spectrometry to determine phthalate esters in beverages
Cao X, et al.
Journal of Separation Science, 37(24), 3677-3683 (2014)
Chenchen Chi et al.
Journal of environmental sciences (China), 55, 137-145 (2017-05-10)
Based on previous research, the sampling and analysis methods for phthalate esters (PAEs) were improved by increasing the sampling flow of indoor air from 1 to 4L/min, shortening the sampling duration from 8 to 2hr. Meanwhile, through the optimization of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service