Skip to Content
Merck
All Photos(1)

Key Documents

61768

Supelco

Lectin, immobilized on Agarose CL-4B from Triticum vulgaris

suitable for affinity chromatography

Synonym(s):

Wheat germ agglutinin, immobilized on agarose CL-4B

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
R$10,685.00

R$10,685.00


Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
5 ML
R$10,685.00

About This Item

UNSPSC Code:
12190000
NACRES:
NA.21

R$10,685.00


Estimated to ship onJune 02, 2025


Request a Bulk Order

technique(s)

affinity chromatography: suitable

Quality Level

capacity

0.1-1.0 mg/mL, packed gel capacity (ovomucoid)

storage temp.

2-8°C

Physical form

50% aqueous suspension in 1 M NaCl

Other Notes

Protein isolation from bordetella pertussis that facilitates entry of calmodulin-sensitive adenylate cyclase into animal cells[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M G Donovan et al.
Biochemistry, 28(20), 8124-8129 (1989-10-03)
Bordetella pertussis, the pathogen responsible for whooping cough, releases a soluble calmodulin-sensitive adenylate cyclase into its culture medium. Several investigators have shown that the partially purified adenylate cyclase is capable of entering animal cells and elevating intracellular cAMP levels [Confer

Articles

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

This page shows how to solve practical problems that may occur when running an affinity chromatography column.

Protocols

This page shows how to purify glycoproteins and polysaccharides by affinity chromatography using Agarose Wheat Germ Lectin.

This page shows how to convert between linear flow and volumetric flow rates in affinity chromatography.

This page shows how to perform column packing and preparation for affinity chromatography when using Tricorn™ or XK columns available from Cytiva.

This page shows how to prepare samples for purification with affinity chromatography.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service