Skip to Content
Merck
All Photos(1)

Documents

60660

Sigma-Aldrich

3-sn-Phosphatidylethanolamine solution

from soybean, ≥97.0% (10 mg phospholipid per ml CHCl3, TLC)

Synonym(s):

L-α-Cephalin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

biological source

soybean

Quality Level

Assay

≥97.0% (10 mg phospholipid per ml CHCl3, TLC)

form

liquid

lipid type

phosphoglycerides

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C40H80NO8P/c1-3-5-7-9-11-13-15-17-18-19-20-21-23-25-27-29-31-33-40(43)49-38(37-48-50(44,45)47-35-34-41)36-46-39(42)32-30-28-26-24-22-16-14-12-10-8-6-4-2/h38H,3-37,41H2,1-2H3,(H,44,45)/t38-/m1/s1

InChI key

NJGIRBISCGPRPF-KXQOOQHDSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miki Shimbo et al.
Brain research, 1491, 34-43 (2012-11-28)
Chondroitin sulfate (CS), the carbohydrate chain of chondroitin sulfate proteoglycans, is involved in neuronal circuit formation during development. CS shows great structural diversity with combination of disaccharide units of different structure (A-, C-, D-, or E-unit). However, whether its structural
Shiro Mawatari et al.
Lipids in health and disease, 11, 161-161 (2012-11-23)
Many disorders with plasmalogen deficiency have been reported. Replenishment or replacement of tissue plasmalogens of these disorders would be beneficial to the patients with these disorders, but effects of dietary plasmalogen on mammals have not been reported. Plasmalogens were purified
Allyson A West et al.
The American journal of clinical nutrition, 97(4), 718-727 (2013-03-01)
Phosphatidylcholine (PC) produced via the S-adenosylmethionine-dependent phosphatidylethanolamine (PE) N-methyltransferase (PEMT) pathway is enriched with docosahexaenoic acid (DHA). DHA plays a critical role in fetal development and is linked to health endpoints in adulthood. It is unknown whether choline, which can
Safia Nathoo et al.
Chemistry and physics of lipids, 167-168, 33-42 (2013-02-07)
Understanding lipid-protein interactions to enhance our knowledge of membrane architecture is a critical step in the development of novel therapeutic measures to respond to the drastic rise of drug resistant microorganisms. Escherichia coli contains a small archetypal inner membrane multidrug
Chiahsin Lin et al.
PloS one, 8(2), e57823-e57823 (2013-03-08)
Given the previously documented importance of lipid concentration and composition in the successful cryopreservation of gorgonian corals, these parameters were assessed in oocytes of five species of scleractinian coral; Platygyra daedalea, Echinopora gemmacea, Echinophyllia aspera, Oxypora lacera and Astreopora expansa.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service