57980
Iodoform
purum, ≥99.0% (AT)
Synonym(s):
Triiodomethane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
CHI3
CAS Number:
Molecular Weight:
393.73
Beilstein:
1697010
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
grade
purum
Assay
≥99.0% (AT)
mp
118-121 °C (lit.)
118-121 °C
density
4.008 g/mL at 25 °C (lit.)
SMILES string
IC(I)I
InChI
1S/CHI3/c2-1(3)4/h1H
InChI key
OKJPEAGHQZHRQV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Ying Xia et al.
Water research, 124, 219-226 (2017-08-02)
Iodide (I
Hervé Gallard et al.
Environmental science & technology, 43(18), 7003-7009 (2009-10-08)
This study shows that iodinated organic compounds can be produced when iodide-containing waters are in contact with manganese oxide birnessite (delta-MnO2) in the pH range of 5-7. In the absence of natural organic matter (NOM), iodide is oxidized to iodate
Eduardo Antunes Bortoluzzi et al.
Oral surgery, oral medicine, oral pathology, oral radiology, and endodontics, 108(4), 628-632 (2009-08-25)
The objective of this study was to evaluate the radiopacity of white Portland cement (PC) associated with bismuth oxide (Bi(2)O(3)), barium sulfate (BaSO(4)), iodoform (CHI(3)), and zirconium oxide (ZrO(2)). White mineral trioxide aggregate (WMTA) and PC without radiopacifier were used
Fumihiro Mizokami et al.
Biological & pharmaceutical bulletin, 35(7), 1048-1053 (2012-07-14)
Iodoform gauze is used in clinical practice for treatment of infected wounds. However, effectiveness and action mechanism of iodoform gauze for removal of necrotic tissue are unknown. We therefore employed case control and biochemical studies in order to clarify the
Anieli Dossa Bergoli et al.
The Journal of clinical pediatric dentistry, 35(2), 191-195 (2010-01-01)
This study investigates the profile of teaching primary tooth pulp therapy practiced by Brazilian dental schools. A multiple-choice questionnaire was sent by e-mail to 191 dental schools in Brazil, addressed to the pediatric dentistry Chairperson. The two-part survey consisting of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service