Skip to Content
Merck
All Photos(2)

Key Documents

55901

Supelco

Ecamsule

analytical standard

Synonym(s):

3,3′-(1,4-Phenylenedimethylidyne)bis[7,7-dimethyl-2-oxo-bicyclo[2.2.1]heptane-1-methanesulfonic acid], Terephthalylidene-3,3′-dicamphor-10,10′-disulfonic acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
R$936.00

R$936.00


Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
R$936.00

About This Item

Empirical Formula (Hill Notation):
C28H34O8S2
CAS Number:
Molecular Weight:
562.69
Beilstein:
8371688
EC Number:
UNSPSC Code:
77101502
NACRES:
NA.24

R$936.00


Estimated to ship onMay 31, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥97.0% (T)
≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

composition

sulfur content, 11.40%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CC1(C)C2CCC1(CS(O)(=O)=O)C(=O)\C2=C/c3ccc(cc3)\C=C4\C5CCC(CS(O)(=O)=O)(C4=O)C5(C)C

InChI

1S/C28H34O8S2/c1-25(2)21-9-11-27(25,15-37(31,32)33)23(29)19(21)13-17-5-7-18(8-6-17)14-20-22-10-12-28(24(20)30,26(22,3)4)16-38(34,35)36/h5-8,13-14,21-22H,9-12,15-16H2,1-4H3,(H,31,32,33)(H,34,35,36)/b19-13-,20-14+

InChI key

HEAHZSUCFKFERC-LRVMPXQBSA-N

General description

Ecamsule is an organic compound added to sunscreens to filter out UVA rays and to protect the skin from adverse effects of ultraviolet light like DNA damage, photo-aging and sunburns.[1]

Application

Ecamsule (ECAM) has been used as a reference standard for observing the photo-stability and photo-sensitizing property of the ecamsule that forms a major ingredient in the production of sunscreen products by laser flash photolysis and steady-state HPLC coupled with mass spectrometry method.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Photo-stability and photo-sensitizing characterization of selected sunscreens? ingredients.
Abid A R, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 332, 241-250 (2017)

Questions

  1. which tempreture should we keep the powder.?

    1 answer
    1. This product may be stored at room temperature.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service