Skip to Content
Merck
All Photos(1)

Key Documents

55769

Supelco

Linolelaidic acid solution

~250 mg/mL in ethanol, analytical standard

Synonym(s):

trans-9,trans-12-Octadecadienoic acid solution, Linoelaidic acid solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H32O2
CAS Number:
Molecular Weight:
280.45
Beilstein:
2210856
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

form

liquid

shelf life

limited shelf life, expiry date on the label

concentration

~250 mg/mL in ethanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

single component solution

functional group

carboxylic acid

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCCCC\C=C\C\C=C\CCCCCCCC(O)=O

InChI

1S/C18H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h6-7,9-10H,2-5,8,11-17H2,1H3,(H,19,20)/b7-6+,10-9+

InChI key

OYHQOLUKZRVURQ-AVQMFFATSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ulaganathan Mabalirajan et al.
Scientific reports, 3, 1349-1349 (2013-02-28)
Airway epithelial injury is the hallmark of various respiratory diseases, but its mechanisms remain poorly understood. While 13-S-hydroxyoctadecadienoic acid (13-S-HODE) is produced in high concentration during mitochondrial degradation in reticulocytes little is known about its role in asthma pathogenesis. Here
Soottawat Benjakul et al.
Food science and technology international = Ciencia y tecnologia de los alimentos internacional, 19(4), 365-376 (2013-06-05)
Antioxidant activity of brown lead (Leucaena leucocephala) seed extracts with and without prior chlorophyll removal was studied in comparison with mimosine. Both extracts showed similar hydroxyl radical (HO(•)) scavenging activity, hydrogen peroxide (H2O2) scavenging activity, singlet oxygen inhibition and hypochlorous
Martijn B Katan
Nederlands tijdschrift voor geneeskunde, 157(22), A6290-A6290 (2013-05-30)
A re-analysis of the Sydney Diet Heart Study (Ramsden et al., BMJ 2013) suggested that a diet high in linoleic acid increased the risk of coronary heart disease. The authors ascribe this to a lack of the omega-3 fatty acid
ACP Journal Club. Review: replacing dietary saturated fatty acids with n-6 polyunsaturated fatty acids does not reduce mortality.
Kathleen Fairfield
Annals of internal medicine, 158(10), JC6-JC6 (2013-05-22)
Saïd Gharby et al.
Natural product communications, 8(1), 25-28 (2013-03-12)
Deforestation is an important matter for the argan forest whose preservation necessitates planting trees. Macroscopic parameters are urgently needed to identify trees presenting good potential as oil producers. This study demonstrates that argan oil produced from kernels of apiculate fruit

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service