Skip to Content
Merck
All Photos(2)

Documents

55669

Supelco

4-Acetamidoantipyrine

analytical standard

Synonym(s):

4-Acetylaminophenazone, N-(2,3-Dihydro-1,5-dimethyl-3-oxo-2-phenyl-1H-pyrazol-4-yl)acetamide, N-Antipyrinylacetamide, NSC 331807

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H15N3O2
CAS Number:
Molecular Weight:
245.28
Beilstein:
234585
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.0% water

mp

200-203 °C (lit.)

application(s)

pharmaceutical

format

neat

SMILES string

CN1N(C(=O)C(NC(C)=O)=C1C)c2ccccc2

InChI

1S/C13H15N3O2/c1-9-12(14-10(2)17)13(18)16(15(9)3)11-7-5-4-6-8-11/h4-8H,1-3H3,(H,14,17)

InChI key

OIAGWXKSCXPNNZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

María Jesús Martínez Bueno et al.
Analytical chemistry, 79(24), 9372-9384 (2007-11-16)
This paper describes an enhanced liquid chromatography-mass spectrometry (LC-MS) strategy for the analysis of a selected group of 56 organic pollutants in wastewater. This group comprises 38 pharmaceuticals and 10 of their most frequent metabolites, 6 pesticides, and 2 disinfectants.
Lu-Qi Zhang et al.
Journal of mass spectrometry : JMS, 55(3), e4497-e4497 (2020-01-10)
A multiclass and multiresidue method for screening veterinary drugs and pesticides in infant formula was developed and validated using ultrahigh-performance liquid chromatography coupled to Quadrupole-Orbitrap high-resolution mass spectrometry (UHPLC-HRMS). A total of 49 veterinary drugs and pesticides investigated belong to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service