Skip to Content
Merck
All Photos(1)

Documents

55667

Supelco

6,10-Dimethyl-5,9-undecadien-2-one, mixture of isomers

analytical standard

Synonym(s):

Mixture of Nerylacetone and Geranylacetone, Geranylacetone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)=CHCH2CH2COCH3
CAS Number:
Molecular Weight:
194.31
Beilstein:
1722277
EC Number:
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.465-1.469

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

C\C(C)=C\CC\C(C)=C\CCC(C)=O

InChI

1S/C13H22O/c1-11(2)7-5-8-12(3)9-6-10-13(4)14/h7,9H,5-6,8,10H2,1-4H3/b12-9+

InChI key

HNZUNIKWNYHEJJ-FMIVXFBMSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jisun Hj Lee et al.
Journal of the science of food and agriculture, 101(1), 307-314 (2020-07-06)
Polyphenol oxidase (PPO) and peroxidase (POD) are key enzymes associated with shelf life and defense responses. Thus, the activity of PPO and POD enzymes is usually assessed to check the quality of food samples and to understand the physiological responses
Merybeth F Triana et al.
PloS one, 15(4), e0231689-e0231689 (2020-04-17)
The giant sugarcane borer Telchin licus (Drury, 1773) (Lepidoptera: Castniidae) is a day-flying moth pest of sugarcane, pineapples and bananas. To better understand the chemical communication in this species, we examined the morphology of its olfactory system and the chemical
Souleymane Diallo et al.
Frontiers in cellular neuroscience, 14, 137-137 (2020-06-26)
Insects that transmit many of the world's deadliest animal diseases, for instance trypanosomosis, find their suitable hosts and avoid non-preferred hosts mostly through olfactory cues. The waterbuck repellent blend (WRB) comprising geranylacetone, guaiacol, pentanoic acid, and δ-octalactone derived from waterbuck
Hong Jiang et al.
The New phytologist, 227(4), 1213-1221 (2020-04-28)
In Orchidaceae, pollination is mostly animal-mediated, and one-third of species have evolved a deceptive pollination mechanism without rewards. Cypripedium is a representative lineage of nonrewarding orchids restricted to temperate regions. Cypripedium subtropicum flowers are pollinated by hoverflies and have hairy
Hee Ju Yoo et al.
Antioxidants (Basel, Switzerland), 9(1) (2019-12-28)
Light is a major environmental factor affecting the regulation of secondary metabolites, such as pigments and flavor. The Solanaceae plant family has diverse patterns of fruit metabolisms that serve as suitable models to understand the molecular basis of its regulation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service