Skip to Content
Merck
All Photos(1)

Documents

55417

Supelco

1,2-Propanediol

analytical reference material

Synonym(s):

Propylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2OH
CAS Number:
Molecular Weight:
76.09
Beilstein:
1340498
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical reference material

Quality Level

vapor density

2.62 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)

Assay

≥98.5% (qNMR)
≥99.5% (GC)

autoignition temp.

779 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

12.5 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

refractive index

n20/D 1.432 (lit.)
n20/D 1.432-1.434

bp

187 °C (lit.)

mp

−60 °C (lit.)

density

1.036 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
forensics and toxicology
personal care
pharmaceutical (small molecule)

format

neat

SMILES string

CC(O)CO

InChI

1S/C3H8O2/c1-3(5)2-4/h3-5H,2H2,1H3

InChI key

DNIAPMSPPWPWGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

217.4 °F - closed cup

Flash Point(C)

103 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J S LaKind et al.
Critical reviews in toxicology, 29(4), 331-365 (1999-08-18)
The purpose of this article is to review and interpret the scientific literature on the mammalian toxicity of ethylene glycol (EG) and propylene glycol (PG), with the goal of comparing the toxicity of the two chemicals. This type of review
Janina Seubert et al.
PloS one, 9(5), e98347-e98347 (2014-05-31)
Scented cosmetic products are used across cultures as a way to favorably influence one's appearance. While crossmodal effects of odor valence on perceived attractiveness of facial features have been demonstrated experimentally, it is unknown whether they represent a phenomenon specific
J O Funk et al.
Contact dermatitis, 31(4), 236-241 (1994-10-01)
Evaluation of dermatitis associated with propylene glycol application or ingestion remains a challenge. The research dealing with skin reactions to propylene glycol is revisited and new aspects for future research are outlined. Based on literature review and our own observations
Li Wang et al.
Hepatology (Baltimore, Md.), 61(2), 486-496 (2014-09-10)
Defective autophagy is implicated in the pathogenesis of nonalcoholic fatty liver diseases (NAFLD) through poorly defined mechanisms. Cardiolipin is a mitochondrial phospholipid required for bioenergetics and mitophagy from yeast to mammals. Here we investigated a role for ALCAT1 in the
Holger Lessmann et al.
Contact dermatitis, 53(5), 247-259 (2005-11-15)
In the several publications reviewed in this article, propylene glycol (PG; 1,2-propylene glycol) is described as a very weak contact sensitizer, if at all. However, particular exposures to PG-containing products might be associated with an elevated risk of sensitization. To

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service