Skip to Content
Merck
All Photos(1)

Documents

52851

Supelco

N-Sebacoylglycine

analytical standard

Synonym(s):

10-[(Carboxymethyl)amino]-10-oxodecanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H21NO5
CAS Number:
Molecular Weight:
259.30
Beilstein:
2278556
UNSPSC Code:
12352200
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

format

neat

storage temp.

2-8°C

SMILES string

OC(CNC(CCCCCCCCC(O)=O)=O)=O

InChI

1S/C12H21NO5/c14-10(13-9-12(17)18)7-5-3-1-2-4-6-8-11(15)16/h1-9H2,(H,13,14)(H,15,16)(H,17,18)

InChI key

VQSDLNWYRNYTJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service