528021
Dimethyl sulfide
Synonym(s):
DMS, Methyl sulfide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3)2S
CAS Number:
Molecular Weight:
62.13
Beilstein:
1696847
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Recommended Products
vapor density
2.1 (vs air)
vapor pressure
26.24 psi ( 55 °C)
7.79 psi ( 20 °C)
Assay
≥99%
autoignition temp.
402 °F
expl. lim.
19.7 %
availability
available only in EU
refractive index
n20/D 1.435 (lit.)
bp
38 °C (lit.)
mp
−98 °C (lit.)
solubility
water: soluble 7.28 g/L at 20 °C
density
0.846 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
CSC
InChI
1S/C2H6S/c1-3-2/h1-2H3
InChI key
QMMFVYPAHWMCMS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Dimethyl sulfide (DMS) is a biogenic sulfur compound that is released from the ocean to the atmosphere. It forms the main component of the global sulfur cycle. The thermal decomposition behavior of dimethyl sulfide in gas phase has been investigated in a static system at 681-723K. The mechanism of its photochemical oxidation in atmosphere has been described. The rate constants of the gas phase reaction between OH radicals and DMS have been determined by relative rate method and flash photolysis. The differential Raman cross section, σDMS at different excitation wavelengths has been measured. The adsorption of DMS on activated carbons (ACs) has been reported to take place by hydrogen bonding.
Application
Dimethyl sulfide may be used to prepare methanethiol by thiolation reaction with hydrogen sulfide (H2S) in the presence of tungsten-zirconia (WO3/ZrO2) catalyst.
Legal Information
Product of Arkema
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
-32.8 °F - closed cup
Flash Point(C)
-36 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Adsorption of dimethyl sulfide vapors by activated carbons.
Goyal M, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 322(1), 164-169 (2008)
The reaction of OH radicals with dimethyl sulfide.
Wallington TJ, et al.
International Journal of Chemical Kinetics, 18(8), 837-846 (1986)
Thiolation of dimethyl sulfide to methanethiol over WO3/ZrO2 catalysts.
Chen S, et al.
J. Mol. Catal. A: Chem., 365, 60-65 (2012)
The pyrolysis of dimethyl sulfide, kinetics and mechanism.
Shum LGS and Benson SW.
International Journal of Chemical Kinetics, 17(7), 749-761 (1985)
Differential Raman cross section of dimethyl sulfide.
Barletta RE and Roe CH.
Journal of Raman Spectroscopy, 42(1), 117-121 (2011)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service