Skip to Content
Merck
All Photos(1)

Key Documents

52464

Supelco

trans-2-Octenal

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
R$415.00

R$415.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

Select a Size

Change View
1 ML
R$415.00

About This Item

Linear Formula:
CH3(CH2)4CH=CHCHO
CAS Number:
Molecular Weight:
126.20
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

R$415.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

grade

analytical standard

vapor density

>1 (vs air)

Assay

≥96.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

refractive index

n20/D 1.45 (lit.)
n20/D 1.450-1.455

bp

84-86 °C/19 mmHg (lit.)

density

0.846 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

[H]C(=O)C(\[H])=C(/[H])CCCCC

InChI

1S/C8H14O/c1-2-3-4-5-6-7-8-9/h6-8H,2-5H2,1H3/b7-6+

InChI key

LVBXEMGDVWVTGY-VOTSOKGWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1,N6-etheno-2'-deoxyadenosine adducts from trans, trans-2,4-decadienal and trans-2-octenal.
V M Carvalho et al.
Advances in experimental medicine and biology, 500, 229-232 (2002-01-05)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service