Skip to Content
Merck
All Photos(1)

Key Documents

50334

Supelco

Tungsten Standard for ICP

TraceCERT®, 1 g/L W in nitric acid and hydrofluoric acid (nominal concentration)

Synonym(s):

Tungsten ICP Calibration Standard

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
R$1,598.00

R$1,598.00


Available to ship onApril 07, 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
R$1,598.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

R$1,598.00


Available to ship onApril 07, 2025Details


Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

concentration

1 g/L W in nitric acid and hydrofluoric acid (nominal concentration)

technique(s)

ICP: suitable

application(s)

industrial qc
pharmaceutical

format

single component solution

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty, date of expiry, and detailed information about trace impurities.

Application

Tungsten Standard for ICP has been used as a standard stock solution in the determination of tungsten concentration in bulk drugs and intermediates using high performance liquid chromatography combined with inductively coupled plasma mass spectrometry method (HPLC-ICP-MS).[1]

Analysis Note

1 g/L W in 5% nitric acid and 2% hydrofluoric acid, prepared with high purity W metal, HNO3, HF, and water

Other Notes

For a complete product listing of our TraceCERT® range of CRMs for ICP and AAS, technical information and example certificates, please visit our ICP & AAS standards website

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The determination of tungsten, molybdenum, and phosphorus oxyanions by high performance liquid chromatography inductively coupled plasma mass spectrometry
Wang T, et al.
Talanta, 72, 1828-1832 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service