Skip to Content
Merck
All Photos(1)

Key Documents

46968

Supelco

Tolbutamide

VETRANAL®, analytical standard

Synonym(s):

1-Butyl-3-(4-methylphenylsulfonyl)urea, N-[(Butylamino)carbonyl]-4-methylbenzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18N2O3S
CAS Number:
Molecular Weight:
270.35
Beilstein:
1984428
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

CCCCNC(=O)NS(=O)(=O)c1ccc(C)cc1

InChI

1S/C12H18N2O3S/c1-3-4-9-13-12(15)14-18(16,17)11-7-5-10(2)6-8-11/h5-8H,3-4,9H2,1-2H3,(H2,13,14,15)

InChI key

JLRGJRBPOGGCBT-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Anti-diabetic agent. Metabolized by CYP2C9 (tolbutamide hydroxylase).

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Kevin S C Hamming et al.
Diabetes, 59(7), 1686-1693 (2010-04-24)
The sodium-calcium exchanger isoform 1 (NCX1) regulates cytoplasmic calcium (Ca(2+)(c)) required for insulin secretion in beta-cells. NCX1 is alternatively spliced, resulting in the expression of splice variants in different tissues such as NCX1.3 and -1.7 in beta-cells. As pharmacological inhibitors
Yasinalli Tamboli et al.
Bioorganic & medicinal chemistry letters, 22(11), 3810-3815 (2012-05-09)
We describe a new class of NO-donor hypoglycemic products obtained by joining tolbutamide, a typical hypoglycemic sulfonylurea, with a NO-donor moiety through a hard link. As NO-donors we chose either furoxan (1,2,5-oxadiazole 2-oxide) derivatives or the classical nitrooxy function. A
Claire E Hills et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 30(3), 575-586 (2012-07-27)
The extracellular calcium-sensing receptor (CaR) is expressed in pancreatic β-cells where it is thought to facilitate cell-to-cell communication and augment insulin secretion. However, it is unknown how CaR activation improves β-cell function. Immunocytochemistry and western blotting confirmed the expression of
Ruurdtje Hoekstra et al.
Drug metabolism and disposition: the biological fate of chemicals, 41(3), 562-567 (2012-12-15)
The human liver cell line HepaRG has been recognized as a promising source for in vitro testing of metabolism and toxicity of compounds. However, currently the hepatic differentiation of these cells relies on exposure to dimethylsulfoxide (DMSO), which, as a
Genetic factors influencing the metabolism of tolbutamide.
D J Back et al.
Pharmacology & therapeutics, 44(2), 147-155 (1989-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service