46732
Salbutamol hemisulfate salt
VETRANAL®, analytical standard
Synonym(s):
α1-[(tert-Butylamino)methyl]-4-hydroxy-1,3-benzenedimethanol hemisulfate salt, Albuterol sulfate
About This Item
Recommended Products
grade
analytical standard
product line
VETRANAL®
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
clinical
forensics and toxicology
pharmaceutical (small molecule)
format
neat
SMILES string
OS(O)(=O)=O.CC(C)(C)NCC(O)c1ccc(O)c(CO)c1.CC(C)(C)NCC(O)c2ccc(O)c(CO)c2
InChI
1S/2C13H21NO3.H2O4S/c2*1-13(2,3)14-7-12(17)9-4-5-11(16)10(6-9)8-15;1-5(2,3)4/h2*4-6,12,14-17H,7-8H2,1-3H3;(H2,1,2,3,4)
InChI key
BNPSSFBOAGDEEL-UHFFFAOYSA-N
Gene Information
human ... ADRB2(154)
Looking for similar products? Visit Product Comparison Guide
General description
Application
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - Repr. 2 - Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service