Skip to Content
Merck
All Photos(1)

Key Documents

46367

Supelco

2,4,5-T solution

100 μg/mL in acetonitrile, PESTANAL®, analytical standard

Synonym(s):

2,4,5-Trichlorophenoxyacetic acid solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3C6H2OCH2CO2H
CAS Number:
Molecular Weight:
255.48
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

single component solution

storage temp.

2-8°C

SMILES string

OC(=O)COc1cc(Cl)c(Cl)cc1Cl

InChI

1S/C8H5Cl3O3/c9-4-1-6(11)7(2-5(4)10)14-3-8(12)13/h1-2H,3H2,(H,12,13)

InChI key

SMYMJHWAQXWPDB-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y R Wang et al.
Water research, 45(13), 3883-3889 (2011-05-10)
A novel electrochemically enhanced advanced oxidation process for the destruction of organic contaminants in aqueous solution is reported in this study. The process involves the use of an iron (Fe) sheet as sacrificial anode and a graphite bar as cathode.
Maria P Yurkova et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 12(4), 684-689 (2012-10-23)
Photodegradation of herbicide 2,4,5-trichlorophenoxyacetic acid (2,4,5-T) in aqueous solution was investigated by stationary (254 nm) and nanosecond laser (266 nm) flash photolysis techniques. It was shown that in the primary photochemical step both photoionization (which generates a hydrated electron-radical cation
Alvin L Young et al.
Military medicine, 176(7 Suppl), 29-34 (2011-09-16)
Serum dioxin studies of Vietnam (VN) veterans, military historical records of tactical herbicide use in Vietnam, and the compelling evidence of the photodegradation of 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD) and other aspects of environmental fate and low bioavailability of TCDD are consistent with
Anh Duc Ngo et al.
European journal of epidemiology, 25(1), 37-44 (2009-11-07)
The objective of this study is to conduct a meta-analysis of published and unpublished studies that examine the association between Agent Orange (AO) exposure and the risk of spina bifida. Relevant studies were identified through a computerized literature search of
The current level of dioxin pollution in the area of large-scale spraying of Agent Orange in Vietnam.
E S Brodsky et al.
Doklady biological sciences : proceedings of the Academy of Sciences of the USSR, Biological sciences sections, 429, 526-530 (2010-02-23)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service