Skip to Content
Merck
All Photos(2)

Documents

45704

Supelco

Vernolat

PESTANAL®, analytical standard

Synonym(s):

S-Propyl dipropylthiocarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H21NOS
CAS Number:
Molecular Weight:
203.34
Beilstein:
1767659
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCCSC(=O)N(CCC)CCC

InChI

1S/C10H21NOS/c1-4-7-11(8-5-2)10(12)13-9-6-3/h4-9H2,1-3H3

InChI key

OKUGPJPKMAEJOE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Vernolat is an herbicide, also known as S-Propyl dipropylthiocarbamate. It is a thiocarbamate compound that is also used as a general pesticide. It is commonly used to kill weeds in soybeans, peanuts and sweet potatoes .

Biochem/physiol Actions

Vernolat has been shown to inhibit aldehyde dehydrogenase (ALDH) in vivo . It is genotoxic by causing chromosomal changes such as chromatid exchanges and chromosomal aberrations in vitro .

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Pintér et al.
Annali dell'Istituto superiore di sanita, 25(4), 577-582 (1989-01-01)
Twenty-two herbicides were studied in 67 tests for induction of DNA damage, gene mutation and chromosomal changes in vitro and in vivo. Triazine and urea-type herbicides were found to be inactive in all but one test. Of 4 thiocarbamates, molinate
G B Quistad et al.
Life sciences, 55(20), 1537-1544 (1994-01-01)
The herbicide S-ethyl N,N-dipropylthiocarbamate (EPTC) and three of its candidate metabolites (the sulfoxide, N-depropyl and S-methyl derivatives) inhibit mitochondrial low-Km aldehyde dehydrogenase (ALDH) in liver by 56 to 82% 2 hr after these thiocarbamates are administered intraperitoneally (ip) to mice
[Determination of maximum permissible exposure level for the herbicide vernam in the air of a working zone].
V N Karpenko et al.
Gigiena truda i professional'nye zabolevaniia, (9)(9), 49-50 (1983-09-01)
A Pintér et al.
Mutation research, 242(4), 279-283 (1990-12-01)
Three thiocarbamate herbicides, butylate (S-ethyl-diisobutylthiocarbamate), vernolate (S-propyl dipropylthiocarbamate) and molinate (S-ethyl-N,N-hexamethylenethiocarbamate) were assayed for cytogenetic effect in the mouse bone marrow micronucleus test. Butylate was inactive in bone marrow, vernolate caused a marginal increase in the incidence of micronucleated polychromatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service