Skip to Content
Merck
All Photos(1)

Key Documents

45615

Supelco

Perthan

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H20Cl2
CAS Number:
Molecular Weight:
307.26
Beilstein:
2054366
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCc1ccc(cc1)C(C(Cl)Cl)c2ccc(CC)cc2

InChI

1S/C18H20Cl2/c1-3-13-5-9-15(10-6-13)17(18(19)20)16-11-7-14(4-2)8-12-16/h5-12,17-18H,3-4H2,1-2H3

InChI key

QFMDFTQOJHFVNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qingxiang Zhou et al.
Journal of chromatography. A, 1147(1), 10-16 (2007-03-10)
TiO(2) nanotubes, novel nanomaterials synthesized from hydrothermal treatment, were investigated for being used as a new solid-phase extraction adsorbent with o,p'-DDT, [1,1,1-trichloro-2-(o-chlorophenyl)-2-(p-chlorophenyl)ethane, p,p'-DDT, [1,1,1-trichloro-2,2-bis(4-chlorophenyl)ethane] and its principle metabolites p,p'-DDD, [1,1-dichloro-2,2-bis(4-chlorophenyl)ethane] and p,p'-DDE [1,1-(2,2-dichloro-ethanylidene)-bis(4-chlorobenzene)] as the target analytes. Several factors such
Anna Axmon et al.
Chemosphere, 70(9), 1620-1628 (2007-09-12)
To investigate a possible time trend in serum concentrations of persistent organochlorine pollutants (POPs), representative samples of the young (median age 18 years) Swedish male population were investigated in the years 2000 and 2004. Due to their low age, these
M D Reuber
Tumori, 66(3), 277-283 (1980-06-30)
B6C3F1 female mice fed an average dose of 3,000 ppm of perthane for 2 years developed a highly significant incidence of hepatocellular carcinomas of the liver. Male mice fed 2,500 ppm perthane had a slight increase of carcinoma of the
O,P'DDD IN THE TREATMENT OF ADVANCED MAMMARY CARCINOMA.
S WEISENFELD et al.
Cancer, 17, 1258-1266 (1964-10-01)
V W Burse et al.
Journal of analytical toxicology, 14(3), 143-145 (1990-05-01)
Four compounds--2,2', 3,4'-tetrachlorobiphenyl, decachlorodiphenylether, 1,1-dichloro-2,2-bis(p-ethylphenyl)ethane, and dichlorobenzophenone--are recommended for monitoring the within-sample behavior of an analytical method that quantifies chlorinated pesticides and polychlorinated biphenyls (such as Aroclor 1260) in serum using packed column gas chromatography with electron capture detection. Percent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service