Skip to Content
Merck
All Photos(1)

Key Documents

45494

Supelco

Fenuron

PESTANAL®, analytical standard

Synonym(s):

1,1-Dimethyl-3-phenylurea, Fenuron

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12N2O
CAS Number:
Molecular Weight:
164.20
Beilstein:
2208535
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CN(C)C(=O)Nc1ccccc1

InChI

1S/C9H12N2O/c1-11(2)9(12)10-8-6-4-3-5-7-8/h3-7H,1-2H3,(H,10,12)

InChI key

XXOYNJXVWVNOOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yen Hai Dao et al.
Water research, 45(11), 3309-3317 (2011-04-26)
The relative rates of degradation of three hydroxyl radical probe compounds (atrazine, fenuron and parachlorobenzoic acid (pCBA)) by Fe(III)/H(2)O(2) (pH = 2.85), Fe(III)NTA/H(2)O(2) (neutral pH), Fe(II)/O(2), Fe(II)NTA/O(2), Fe(II)/H(2)O(2) and Fe(II)NTA/H(2)O(2) (neutral pH) have been investigated using the competitive kinetic method.
C Richard et al.
Environmental science & technology, 38(7), 2052-2057 (2004-04-29)
The IHSS soil humic acid (HA) standard and two HAs from soils of very different origin (Chernozem and Ranker) were fractionated by tandem size-exclusion chromatography-polyacrylamide gel electrophoresis. From each HA, three fractions with different molecular sizes (MSs) and electrophoretic mobilities
[Embryotropic effect of fenuron].
L T Voloshina
Vrachebnoe delo, (2)(2), 103-105 (1985-02-01)
Humic substances as natural photoinducers degrading pesticides in the environment.
O E Trubetskaya et al.
Doklady biological sciences : proceedings of the Academy of Sciences of the USSR, Biological sciences sections, 406, 94-96 (2006-04-01)
[Experience in the field of gas chromatographic herbicide residue analysis illustrated by using phenmedipham and fenuron as examples].
G Schumann
Zeitschrift fur die gesamte Hygiene und ihre Grenzgebiete, 25(12), 926-928 (1979-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service