Skip to Content
Merck
All Photos(1)

Key Documents

45328

Supelco

ANTU

PESTANAL®, analytical standard

Synonym(s):

N-(1-Naphthyl)thiourea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10N2S
CAS Number:
Molecular Weight:
202.28
Beilstein:
778118
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

NC(=S)Nc1cccc2ccccc12

InChI

1S/C11H10N2S/c12-11(14)13-10-7-3-5-8-4-1-2-6-9(8)10/h1-7H,(H3,12,13,14)

InChI key

PIVQQUNOTICCSA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Carc. 2

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

José A Murillo Pulgarín et al.
Journal of agricultural and food chemistry, 53(17), 6609-6615 (2005-08-18)
A novel chemiluminescence method for the determination of antu has been developed based on the reaction between potassium permanganate in acid medium with this rat-poison in the presence of formaldehyde as an emission enhancer. The main feature of the system
E Sipahi et al.
Pharmacological research, 33(6), 375-378 (1996-06-01)
Alpha-naphthylthiourea when injected intraperitoneally to rats (10 mg kg-1 i.p.) produced lung oedema as indicated by an increase in lung weight/body ratio and pleural effusion reaching a maximum within 4 hours. Prior intravenous single bolus injection of endothelin-1 elicited a
Mustafa Comert et al.
European journal of pharmacology, 511(2-3), 183-189 (2005-03-29)
This study was designed to investigate the possible participation of morphine in pulmonary oedema induced by alpha-naphthylthiourea (ANTU), which is a well-known noxious chemical agent in the lung. Injection of ANTU (15 mg/kg i.p.) produced pulmonary oedema as indicated by
B P Guery et al.
Journal of applied physiology (Bethesda, Md. : 1985), 85(3), 842-848 (1998-09-08)
Several methodologies have been developed to assess alveolocapillary membrane permeability in acute lung injury. The purpose of this study was to determine the reliability of FITC-dextran compared with radioactive tracers to assess lung permeability alterations. After intraperitoneal administration of alpha-naphthylthiourea
Yong Chen et al.
Organic & biomolecular chemistry, 9(15), 5530-5534 (2011-06-23)
A naphthylthiourea-modified cyclodextrin (1) and its urea derivative (2) were synthesized, and their fluorescence behaviors in the presence of various metal ions were investigated. Significantly, 1 showed a highly sensitive and selective fluorescence sensing ability for Hg(2+) over other metal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service