Skip to Content
Merck
All Photos(1)

Key Documents

43937

Sigma-Aldrich

Di(β-D-xylopyranosyl)amine

≥97.0% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19NO8
CAS Number:
Molecular Weight:
281.26
Beilstein:
3908092
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

Assay

≥97.0% (TLC)

impurities

≤2% water

solubility

H2O: 50 mg/mL, clear, yellow

storage temp.

2-8°C

SMILES string

O[C@@H]1CO[C@@H](N[C@@H]2OC[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O

InChI

1S/C10H19NO8/c12-3-1-18-9(7(16)5(3)14)11-10-8(17)6(15)4(13)2-19-10/h3-17H,1-2H2/t3-,4-,5+,6+,7-,8-,9-,10-/m1/s1

InChI key

WDQLRJPDYLYTMJ-IJNKSVJISA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Di-β-D-xylopyranosylamine is a diglycosylamine generated from the transglycosylation of β-D-xylopyranosylamine.
Structure and rearrangement reactions of some diglycosylamines[1]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K. Linek et al.
Carbohydrate Research, 164, 195-195 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service