Skip to Content
Merck
All Photos(2)

Key Documents

43805

Supelco

Omeprazole 5-carboxylic acid disodium salt

analytical standard

Synonym(s):

4-Methoxy-6-{[(5-methoxy-1H-benzimidazol-2-yl)sulfinyl]methyl}-5-methyl-3-pyridinecarboxylic acid disodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$9,372.00

R$9,372.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
R$9,372.00

About This Item

Empirical Formula (Hill Notation):
C17H15N3Na2O5S
CAS Number:
Molecular Weight:
419.36
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

R$9,372.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[S](=O)(Cc6ncc(c(c6C)OC)C(=O)[O-])c4[nH]c5c(n4)cc(cc5)OC.[S](=O)(Cc3ncc(c(c3C)OC)C(=O)[O-])c1[nH]c2c(n1)cc(cc2)OC

InChI

1S/2C17H17N3O5S.2Na/c2*1-9-14(18-7-11(16(21)22)15(9)25-3)8-26(23)17-19-12-5-4-10(24-2)6-13(12)20-17;;/h2*4-7H,8H2,1-3H3,(H,19,20)(H,21,22);;/q;;2*+1/p-2

InChI key

TUCNPUIUNPRUHQ-UHFFFAOYSA-L

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Renberg et al.
Drug metabolism and disposition: the biological fate of chemicals, 17(1), 69-76 (1989-01-01)
The excretion and metabolism of [14C]omeprazole given orally as a suspension was studied in 10 healthy male subjects. An average of 79% of the dose was recovered in the urine in 96 hr, with most of the radioactivity (76% of
K Kobayashi et al.
Journal of pharmaceutical and biomedical analysis, 12(6), 839-844 (1994-06-01)
A high-performance liquid chromatography assay was developed to measure the enzymatic activities of the 5-hydroxylation and sulphoxidation of omeprazole, a proton pump inhibitor, in human liver microsomes. The preliminary study was also undertaken to assess the assay's applicability for the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service