Skip to Content
Merck
All Photos(2)

Key Documents

42635

Supelco

Ginsenoside Rb3

analytical standard

Synonym(s):

(3β,12β)-3-[(2-O-β-D-Glucopyranosyl-β-D-glucopyranosyl)oxy]-12-hydroxydammar-24-en-20-yl 6-O-β-D-xylopyranosyl-β-D-glucopyranoside, Gypenoside IV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H90O22
CAS Number:
Molecular Weight:
1079.27
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

C\C(C)=C/CC[C@](C)(O[C@@H]1O[C@H](CO[C@@H]2OC[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@H](O)[C@H]1O)[C@H]3CC[C@]4(C)[C@@H]3[C@H](O)C[C@@H]5[C@@]6(C)CC[C@H](O[C@@H]7O[C@H](CO)[C@@H](O)[C@H](O)[C@H]7O[C@@H]8O[C@H](CO)[C@@H](O)[C@H](O)[C@H]8O)C(C)(C)[C@@H]6CC[C@@]45C

InChI

1S/C53H90O22/c1-23(2)10-9-14-53(8,75-47-43(67)39(63)37(61)29(72-47)22-69-45-41(65)34(58)26(57)21-68-45)24-11-16-52(7)33(24)25(56)18-31-50(5)15-13-32(49(3,4)30(50)12-17-51(31,52)6)73-48-44(40(64)36(60)28(20-55)71-48)74-46-42(66)38(62)35(59)27(19-54)70-46/h10,24-48,54-67H,9,11-22H2,1-8H3/t24-,25+,26+,27+,28+,29+,30-,31+,32-,33-,34-,35+,36+,37+,38-,39-,40-,41+,42+,43+,44+,45-,46-,47-,48-,50-,51+,52+,53-/m0/s1

InChI key

NODILNFGTFIURN-USYOXQFSSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qi Tao Bu et al.
Medicinal chemistry (Shariqah (United Arab Emirates)), 8(5), 934-941 (2012-06-30)
As one of the main active component of protopanaxdiol type ginsenosides, ginsenoside Rb(3) is rarely reported in the treatment of diabetes. The anti-diabetic activity of ginsenoside Rb(3) was investigated in a model of alloxan-induced diabetic mice in the present study.
Shan Jiang et al.
Journal of ethnopharmacology, 133(2), 914-916 (2010-10-26)
To investigate the effect of ginsenoside Rb(3) on synaptic transmission after oxygen-glucose deprivation in vitro. The population spike (PS) was recorded in the stratum pyramidale of mouse hippocampal slices using extracellular recordings. Ginsenoside Rb(3) depressed the basal synaptic transmission, which
Kejiang He et al.
Chemical & pharmaceutical bulletin, 53(2), 177-179 (2005-02-03)
A dammarane glycoside, designated compound Mx (C-Mx), was isolated from the hydrolysate of 20(S)-protopanaxadiol type ginsenosides containing G-Rb(3) from Panax notoginseng leaves with crude snailase. Its chemical structure was elucidated to be 20-O-beta-D-xylopyranosyl(1-->6)-beta-D-glucopyranosyl-20(S)-protopanaxadiol on the basis of spectral analysis. Its
Hong-Mei Shen et al.
Zhongguo ying yong sheng li xue za zhi = Zhongguo yingyong shenglixue zazhi = Chinese journal of applied physiology, 22(1), 31-34 (2006-02-01)
To observe protective effects of ginsenoside Rb3 on glutamate excitotoxic injury in cultured hippocampal neurons and involved mechanisms. On cultured rat hippocampal neurons treated with glutamate at toxic concentration, we made the following investigations: by using MTT assay, LDH leakage
Tian Wang et al.
European journal of pharmacology, 636(1-3), 121-125 (2010-04-08)
The present study was designed to evaluate the effect of ginsenoside Rb3 on myocardial injury and heart function impairment induced by isoproterenol in rats. To induce myocardial ischemia, Sprague-Dawley rats were subcutaneously injected with isoproterenol (20mg/kg). Cardiac marker enzymes and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service