Skip to Content
Merck
All Photos(1)

Key Documents

42100

Supelco

Nα-(2,4-Dinitro-5-fluorophenyl)-D-valinamide

for chiral derivatization, LiChropur, ≥98.0%

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
R$8,803.00

R$8,803.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
R$8,803.00

About This Item

Empirical Formula (Hill Notation):
C11H13FN4O5
CAS Number:
Molecular Weight:
300.24
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.22

R$8,803.00


Check Cart for Availability

Request a Bulk Order

grade

derivatization grade ((chiral))
for chiral derivatization

Quality Level

Assay

≥98.0% (sum of enantiomers, HPLC)
≥98.0%

form

solid

optical activity

[α]20/D −14.0±1.5°, c = 2% in acetone

optical purity

enantiomeric ratio: ≥99.5:0.5 (HPLC)

quality

LiChropur

technique(s)

HPLC: suitable

mp

173-177 °C

storage temp.

2-8°C

SMILES string

CC(C)[C@@H](Nc1cc(F)c(cc1[N+]([O-])=O)[N+]([O-])=O)C(N)=O

InChI

1S/C11H13FN4O5/c1-5(2)10(11(13)17)14-7-3-6(12)8(15(18)19)4-9(7)16(20)21/h3-5,10,14H,1-2H3,(H2,13,17)/t10-/m1/s1

InChI key

ZTFFRKNPAXXEBL-SNVBAGLBSA-N

General description

Nα-(2,4-Dinitro-5-fluorophenyl)-D-valinamide [D-FDVA] is a derivatizing agent.[1]

Application

D-FDVA may be used as derivatizing agent for nullifying the possibility of minor compounds to be diastereomer of xenortide A, during HPLC-MS screening.[1]

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gerhard Lang et al.
Journal of natural products, 71(6), 1074-1077 (2008-05-22)
Three new peptides, xenortides A and B and xenematide, were isolated from a culture of the nematode-associated entomopathogenic bacterium Xenorhabdus nematophilus. Their structures were elucidated using NMR, MS, and chemical derivatization methods. Xenortides A and B are the N-phenethylamide and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service