Skip to Content
Merck
All Photos(1)

Key Documents

41712

Millipore

Baird-Parker Agar

NutriSelect® Plus, according to GB 4789.10-2016, powder, final pH 7.0±0.2, pkg of 500 g, non-sterile

Synonym(s):

BPA, Baird Parker Agar Base acc. GB

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
R$291.00

R$291.00


Estimated to ship onMarch 31, 2025


Request a Bulk Order

Select a Size

Change View
500 G
R$291.00

About This Item

UNSPSC Code:
41171606
NACRES:
NA.74

R$291.00


Estimated to ship onMarch 31, 2025


Request a Bulk Order

Agency

according to GB 4789.10-2016

Quality Level

sterility

non-sterile

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 20 g/L
beef extract, 5 g/L
glycine, 12 g/L
lithium chloride (LiCl·6H2O), 5 g/L
sodium pyruvate, 10 g/L
tryptone, 10 g/L
yeast extract, 1 g/L

packaging

pkg of 500 g

manufacturer/tradename

NutriSelect® Plus

final pH

7.0±0.2

application(s)

food and beverages
microbiology

suitability

Staphylococcus spp.

Application

Baird Parker Agar Base is recommended for the isolation and enumeration of coagulase positive Staphylococci from food and other materials.

Preparation Note

Suspend 63 g of the powder in 950 mL of purified water. Mix thoroughly. Heat with frequent agitation and boil to completely dissolve the powder. Sterilize by autoclaving at 15 lbs pressure (121°C) for 15 minutes. Cool to 50°C and aseptically add 50 mL of Egg Yolk Tellurite enrichment (Cat. No.75208). Mix thoroughly but gently and pour into sterile petri plates.

Warning : Lithium Chloride is harmful. Avoid all bodily contact and inhalation of vapours. On contact with skin wash with plenty of water immediately.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service