Skip to Content
Merck
All Photos(1)

Key Documents

41423

Supelco

p-Tolualdehyde

analytical standard

Synonym(s):

4-Methylbenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CHO
CAS Number:
Molecular Weight:
120.15
Beilstein:
385772
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% o-tolualdehyde (GC)
≤0.50% m-tolualdehyde (GC)
≤1.0% water
≤3.0% free carboxylic acid (T)

refractive index

n20/D 1.544-1.548
n20/D 1.545 (lit.)

bp

204-205 °C (lit.)
82-85 °C/11 mmHg (lit.)

density

1.019 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

[H]C(=O)c1ccc(C)cc1

InChI

1S/C8H8O/c1-7-2-4-8(6-9)5-3-7/h2-6H,1H3

InChI key

FXLOVSHXALFLKQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

p-Tolualdehyde (4-Methylbenzaldehyde) is a methylated benzene[1] that belongs to the class of aromatic aldehydes.[2] It is identified as a flavor/aroma component in Greek cotton honey,[3] Brazilian wines,[4] tobacco, etc.[1] 4-Methylbenzaldehyde is also reported to be produced by the members of the genus, Phellinus.[5]

Application

p-Tolualdehyde may be used as an analytical reference standard for the quantification of the analyte in the following:
  • Air samples using high-performance liquid chromatography with UV detection (HPLC-UV).[6]
  • Mango cultivars using gas chromatography (GC) and gas chromatography-mass spectrometry (GC-MS).[7]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flavour compounds of Greek cotton honey
Alissandrakis E, et al.
Journal of the Science of Food and Agriculture, 85(9), 1444-1452 (2005)
Kinetics of gas phase reactions of OH and Cl with aromatic aldehydes
Thiault G, et al.
Physical Chemistry Chemical Physics, 4(11), 2194-2199 (2002)
Analysis of the aroma components in tobacco using combined GC-MS and AMDIS
Wu L, et al.
Analytical Methods : Advancing Methods and Applications, 5(5), 1259-1263 (2013)
Gas-phase absorption cross sections of 24 monocyclic aromatic hydrocarbons in the UV and IR spectral ranges
Etzkorn T, et al.
Atmospheric Environment, 33(4), 525-540 (1999)
Basidiomycetes as new biotechnological tools to generate natural aromatic flavours for the food industry
Lomascolo A, et al.
Trends in Biotechnology, 17(7), 282-289 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service