Skip to Content
Merck
All Photos(1)

Documents

40120

Sigma-Aldrich

Diethoxydimethylsilane

purum, ≥97.0% (GC)

Synonym(s):

Dimethyldiethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2Si(OC2H5)2
CAS Number:
Molecular Weight:
148.28
Beilstein:
1736110
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

purum

Quality Level

Assay

≥97.0% (GC)

form

liquid

color

APHA: ≤25

refractive index

n20/D 1.381 (lit.)

bp

114 °C (lit.)

density

0.865 g/mL at 25 °C (lit.)

SMILES string

CCO[Si](C)(C)OCC

InChI

1S/C6H16O2Si/c1-5-7-9(3,4)8-6-2/h5-6H2,1-4H3

InChI key

YYLGKUPAFFKGRQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Repr. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Zhang et al.
Journal of dentistry, 42(9), 1078-1095 (2014-06-17)
The present study reported a method for preparing a blend of antibacterial quaternary ammonium silanes and quaternary ammonium methacryloxy silane (QAMS) based on the sol-gel reaction between dimethyldiethoxy silane and two trialkoxysilanes, one with an antibacterial quaternary ammonium functionality and
M S Attia et al.
Analytica chimica acta, 835, 56-64 (2014-06-24)
A new method in which a nano optical sensor for diagnosis of different diseases of seminal vesicle and sexual gland was prepared. The working principle of the method depends on the determination of the fructose concentration in semen of different

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service