Skip to Content
Merck
All Photos(1)

Key Documents

38944

Millipore

MRS Broth modified, Vegitone

suitable for microbiology, NutriSelect® Plus

Synonym(s):

DeMan, Rogosa and Sharpe Agar modified, Vegitone, Vegitone MRS Broth modified

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
R$834.00

R$834.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 G
R$834.00

About This Item

UNSPSC Code:
41171607
NACRES:
NA.85

R$834.00


Check Cart for Availability

Request a Bulk Order

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

ammonium citrate, 2 g/L
bromocresol green, 0.04 g/L
captan, 0.004 g/L
dextrose, 20 g/L
dipotassium phosphate, 2 g/L
magnesium sulfate, 0.10 g/L
manganese sulfate, 0.05 g/L
2-phenylethyl alcohol, 3 g/L
proteose peptone (vegetable), 10 g/L
sodium acetate, 5 g/L
yeast extract, 5 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

4.3±0.2 (25 °C)

application(s)

agriculture
bioburden testing
environmental
food and beverages

microbiology

suitability

selective by low pH for Bifidobacterium spp.
selective by low pH for Lactobacillus spp.

Application

This MRS Broth contains plant peptone instead of animal peptone. It is recommended for the isolation and cultivation of Lactobacillus species.

Preparation Note

Suspend 47.2 g in 1000 ml distilled water containing 1 ml polysorbate 80 (TWEEN® 80; Sigma P8074). Boil to dissolve the medium completely and mix thoroughly. Sterilize by autoclaving at 15 lbs pressure (121°C) for 15 minutes. If necessary adjust the pH with glacial acetic acid after sterilization.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Traditional methods are based morphology, staining methods, enzyme reactions (metabolism) and diverse media.

Cost-effective molecular biology method for lactobacilli detection is presented as an alternative to PCR-based methods.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service