Skip to Content
Merck
All Photos(1)

Key Documents

37922

Supelco

2,3,4,5,6-Pentabromoethylbenzene

analytical standard

Synonym(s):

Pentabromoethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H5Br5
CAS Number:
Molecular Weight:
500.65
Beilstein:
3133073
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental
petroleum

format

neat

SMILES string

CCc1c(Br)c(Br)c(Br)c(Br)c1Br

InChI

1S/C8H5Br5/c1-2-3-4(9)6(11)8(13)7(12)5(3)10/h2H2,1H3

InChI key

FIAXCDIQXHJNIX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phoebe J Lewis et al.
Marine pollution bulletin, 159, 111488-111488 (2020-08-02)
While globally distributed throughout the world's ecosystems, there is little baseline information on persistent organic pollutants (POPs) in marine environments in Australia and, more broadly, the Southern Hemisphere. To fill this knowledge gap, we collected baseline information on POPs in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service