Skip to Content
Merck
All Photos(1)

Key Documents

37915

Supelco

Guazatine acetate salt

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
R$997.00

R$997.00


Estimated to ship onMay 22, 2025


Request a Bulk Order

Select a Size

Change View
100 MG
R$997.00

About This Item

CAS Number:
Beilstein:
1995978
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

R$997.00


Estimated to ship onMay 22, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

NC(=N)NCCCCCCCCNCCCCCCCCNC(N)=N

InChI

1S/C18H41N7/c19-17(20)24-15-11-7-3-1-5-9-13-23-14-10-6-2-4-8-12-16-25-18(21)22/h23H,1-16H2,(H4,19,20,24)(H4,21,22,25)

InChI key

RONFGUROBZGJKP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Guazatine acetate salt has been used as an analytical reference standard in the detection and separation of guazatine residues in commercial citrus fruits using high performance liquid chromatography (HPLC) coupled with mass spectrometry (MS/MS).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

LC/MS/MS detection of fungicide guazatine residues for quality assessment of commercial citrus fruit
Scordino M, et al.
European Food Research and Technology, 227, 1339-1347 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service