Skip to Content
Merck
All Photos(2)

Key Documents

37895

Supelco

Carbofuran-3-keto

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13NO4
CAS Number:
Molecular Weight:
235.24
Beilstein:
1581740
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

technique(s)

HPLC: suitable
NMR: suitable
gas chromatography (GC): suitable

mp

175-185 °C

suitability

passes test for identity (NMR)

application(s)

agriculture
environmental

format

neat

storage temp.

−20°C

SMILES string

CNC(=O)Oc1cccc2C(=O)C(C)(C)Oc12

InChI

1S/C12H13NO4/c1-12(2)10(14)7-5-4-6-8(9(7)17-12)16-11(15)13-3/h4-6H,1-3H3,(H,13,15)

InChI key

WXNZYYXXILQTKX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A K Salama
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 33(3), 253-266 (1998-05-30)
Metabolism of carbofuran in pure liquid cultures of A. niger and F. graminearum was investigated. Carbofuran and its metabolites were analyzed by HPLC and TLC. The average recoveries of carbofuran, 3-hydroxycarbofuran, 3-ketocarbofuran, and 3-ketocarbofuran phenol from fungi media were found
J R Miles et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 16(4), 409-417 (1981-01-01)
In a laboratory study, the persistence of carbofuran and its 3-hydroxy- and 3-keto-metabolites was examined separately over 16 wk in sterile and natural organic (muck) and mineral (loam) soils. Carbofuran was relatively persistent in sterile soils; at 8 wk 77%
[Determination of furadan and its metabolites in biomedia by thin-layer chromatography].
V N Os'kina
Gigiena i sanitariia, (7)(7), 45-46 (1982-07-01)
Pei Zhou et al.
Science in China. Series C, Life sciences, 48 Suppl 1, 40-47 (2005-08-11)
The DNA damaging effects of the carbamate pesticide carbofuran and its four metabolites (carbofuranphenol, 3-ketocarbofuran, 3-hydrocarbofuran and nitrosocarbofuran) on mice were evaluated by single cell gel electrophoresis (SCGE) assay and micronucleus test. KM mice were exposed to test compounds with
Kwang-Hyeon Liu et al.
Pest management science, 58(1), 57-62 (2002-02-13)
The pharmacokinetics of furathiocarb were studied in vivo in male Sprague-Dawley rats following dermal treatment. HPLC and post-column derivatization were used for the analysis of furathiocarb and its metabolites (carbofuran, 3-hydroxycarbofuran and 3-ketocarbofuran). Carbofuran and 3-hydroxycarbofuran were detected in plasma

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service