Skip to Content
Merck
All Photos(1)

Key Documents

36846

Supelco

trans-Nonachlor solution

100 μg/mL in acetonitrile, PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H5Cl9
CAS Number:
Molecular Weight:
444.22
Beilstein:
8158568
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

single component solution

storage temp.

2-8°C

SMILES string

Cl[C@@H]1[C@@H](Cl)[C@H]2[C@@H]([C@H]1Cl)[C@]3(Cl)C(Cl)=C(Cl)[C@@]2(Cl)C3(Cl)Cl

InChI

1S/C10H5Cl9/c11-3-1-2(4(12)5(3)13)9(17)7(15)6(14)8(1,16)10(9,18)19/h1-5H/t1-,2+,3+,4-,5-,8-,9+

InChI key

OCHOKXCPKDPNQU-FHPNKJTRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Duk-Hee Lee et al.
Environment international, 47, 115-120 (2012-07-20)
Background exposure to persistent organic pollutants (POPs), lipophilic xenobiotics that accumulate mainly in adipose tissue, has recently emerged as a new risk factor for cardiovascular diseases. This prospective study was performed to evaluate if plasma concentrations of selected POPs predict
Leonard J S Tsuji et al.
Journal of environmental monitoring : JEM, 7(9), 888-898 (2005-08-27)
Abandoned radar line stations in the North American arctic and sub-arctic regions are point sources of contamination, especially for PCBs. Few data exist with respect to human body burden of organochlorines (OCs) in residents of communities located in close proximity
Keri C Hornbuckle et al.
Ambio, 32(6), 406-411 (2003-11-25)
A predictive model for gas-phase PCBs and trans-nonachlor over Lake Michigan has been constructed and the resulting data examined for trends. In this paper, we describe the model results to show how the magnitude and variability of a plume of
Genevieve Bondy et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 42(6), 1015-1027 (2004-04-28)
The chlordane constituent trans-nonachlor and its metabolite oxychlordane are among the most persistent chlordane-related contaminants and are found in tissues and milk from humans ingesting diets high in Arctic marine mammal fat. Although chlordane is no longer registered in North
Duk-Hee Lee et al.
Environmental health perspectives, 118(9), 1235-1242 (2010-05-07)
Low doses of some persistent organic pollutants (POPs) associate cross-sectionally with type 2 diabetes, whereas associations with high POP exposures are inconsistent. We investigated whether several POPs prospectively predict type 2 diabetes within the Coronary Artery Risk Development in Young

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service