Skip to Content
Merck
All Photos(2)

Documents

367648

Sigma-Aldrich

Disperse Blue 14

Dye content 97 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14N2O2
CAS Number:
Molecular Weight:
266.29
Colour Index Number:
61500
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Assay

96.5-103.5%

Quality Level

form

powder

composition

Dye content, 97%

mp

220-222 °C (lit.)

solubility

acetonitrile: 10 mg/mL

λmax

595 nm
644 nm (2nd)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CNc1ccc(NC)c2C(=O)c3ccccc3C(=O)c12

InChI

1S/C16H14N2O2/c1-17-11-7-8-12(18-2)14-13(11)15(19)9-5-3-4-6-10(9)16(14)20/h3-8,17-18H,1-2H3

InChI key

QOSTVEDABRQTSU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service