Skip to Content
Merck
All Photos(1)

Key Documents

36572

Supelco

1,4-Dichloro-2-nitrobenzene

PESTANAL®, analytical standard

Synonym(s):

2,5-Dichloronitrobenzene, Nitro-p-dichlorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3NO2
CAS Number:
Molecular Weight:
192.00
Beilstein:
778109
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

vapor density

6.6 (vs air)

vapor pressure

<0.1 mmHg ( 25 °C)

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

expl. lim.

9.2 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

266-269 °C (lit.)

mp

52-54 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

[O-][N+](=O)c1cc(Cl)ccc1Cl

InChI

1S/C6H3Cl2NO2/c7-4-1-2-5(8)6(3-4)9(10)11/h1-3H

InChI key

RZKKOBGFCAHLCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,4-Dichloro-2-nitrobenzene is a chloronitrobenzene, which can be commonly used in industrial intermediates as an organic dye.[1] It can serve as a substitute for p-dichloroaniline, used in the manufacture of dyes and pigments and it can also find applications as an ultraviolet absorber, which can be useful in stabilizing polyolefins.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

275.0 °F

Flash Point(C)

135 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of N-acetylcysteine conjugate in yellow urine by oral administration of 1, 4-dichloro-2-nitrobenzene in rats
Ohnishi M, et al.
Journal of Health Science, 50, 319-322 (2004)
Carcinogenicity and chronic toxicity of 1, 4-dichloro-2-nitrobenzene in rats and mice by two years feeding
Yamazaki K, et al.
Industrial Health, 44, 230-243 (2006)
Z C Jin et al.
Mutation research, 264(1), 1-5 (1991-09-01)
The plasmid pSK1002 (umuC'-'lacZ) could increase the number of revertants induced by methyl methanesulfonate (MMS) and 4-nitroquinoline-N-oxide (4NQO) in Salmonella typhimurium TA 1535 (his-). The values induced by N-methyl-N'-nitro-N-nitrosoguanidine (MNNG) were not different irrespective of the presence or absence of
Z C Jin et al.
Zhonghua yu fang yi xue za zhi [Chinese journal of preventive medicine], 25(5), 292-295 (1991-09-01)
The plasmid pSK1002 carrying the fused gene umuC'-'lacZ could increase the number of revertants induced by methyl methanesulfonate (MMS) and 4-nitroquinoline-1-oxide (4NQO) in S. typhimurium LT2 TA 1535(his-). Maximum revertants were 4.35 and 3.96 times that of the controls without

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service