Skip to Content
Merck
All Photos(1)

Key Documents

36145

Supelco

MCPB

PESTANAL®, analytical standard

Synonym(s):

4-(4-Chloro-2-methylphenoxy)butyric acid, 4-(4-Chloro-o-tolyloxy)butyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13ClO3
CAS Number:
Molecular Weight:
228.67
Beilstein:
2215202
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Cc1cc(Cl)ccc1OCCCC(O)=O

InChI

1S/C11H13ClO3/c1-8-7-9(12)4-5-10(8)15-6-2-3-11(13)14/h4-5,7H,2-3,6H2,1H3,(H,13,14)

InChI key

LLWADFLAOKUBDR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

MCPB may be used as a reference standard in the determination of MCPB in soil samples using gas chromatography (GC).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid simultaneous determination of residues of MCPA, mecoprop and MCPB in soil by gas chromatography of the pentafluorobenzyl ester.
Cotterill G E.
Journal of Chromatography A, 171, 478-481 (1979)
P Vineis et al.
Scandinavian journal of work, environment & health, 13(1), 9-17 (1987-02-01)
A population-based case-referent study was conducted in an area of northern Italy where rice growing is the predominant agricultural activity and phenoxy herbicides have been used since 1950. Manual rice weeding was formerly performed by a seasonal female working population;
Inhibition of platelet aggregation and thromboxane A2 production by chlorophenoxy acid herbicides.
P Ylitalo et al.
Archives of toxicology. Supplement. = Archiv fur Toxikologie. Supplement, 14, 174-178 (1991-01-01)
S Iu Buslovich et al.
Farmakologiia i toksikologiia, 46(3), 99-101 (1983-05-01)
In acute poisonings with chlorine derivatives of phenoxy acids, the myotonic period of poisoning is marked electromyographically by the manifestations specific and non-specific for myotonia. The characteristic signs involve multiple electrical responses of muscle fibers stimulated by a single excitation
[Use of the orthogonal function method for background suppression in forensic chemical determination of various herbicides in biological material].
T Kh Vergeĭchik et al.
Sudebno-meditsinskaia ekspertiza, 27(3), 47-49 (1984-07-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service