Skip to Content
Merck
All Photos(1)

Documents

34041

Supelco

Orbifloxacin

VETRANAL®, analytical standard

Synonym(s):

1-Cyclopropyl-5,6,8-trifluoro-1,4-dihydro-7-(cis-3,5-dimethyl-1-piperazinyl)-4-oxo-quinoline-3-carboxylic acid, Orbax

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20F3N3O3
CAS Number:
Molecular Weight:
395.38
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

C[C@@H]1CN(C[C@H](C)N1)c2c(F)c(F)c3C(=O)C(=CN(C4CC4)c3c2F)C(O)=O

InChI

1S/C19H20F3N3O3/c1-8-5-24(6-9(2)23-8)17-14(21)13(20)12-16(15(17)22)25(10-3-4-10)7-11(18(12)26)19(27)28/h7-10,23H,3-6H2,1-2H3,(H,27,28)/t8-,9+

InChI key

QIPQASLPWJVQMH-DTORHVGOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Orbifloxacin is an antibacterial fluoroquinolone derivative,for use in small animals such as dogs and cats.

Application

Orbifloxacin may be used as a reference standard in the estimation of orbifloxacin in pharmaceutical formulations using ultraviolet spectrophotometric technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photodegradation products of a new antibacterial fluoroquinolone derivative, orbifloxacin, in aqueous solution.
Morimura T, et al.
Chemical & Pharmaceutical Bulletin, 45(2), 373-377 (1997)
Kirk's Current Veterinary Therapy: Small animal practice (2009)
Development and validation of UV spectrophotometric method for orbifloxacin assay and dissolution studies.
Cazedey LCE and Salgado NRH
Brazilian Journal of Pharmaceutical Sciences , 50(3), 457-465 (2014)
Tao Wang et al.
Arteriosclerosis, thrombosis, and vascular biology, 35(5), 1218-1225 (2015-04-04)
Surgical hindlimb ischemia (HLI) in mice has become a valuable preclinical model to study peripheral arterial disease. We previously identified that the different phenotypic outcomes after HLI across inbred mouse strains is related to a region on the short arm
Cinzia Civitareale et al.
Rapid communications in mass spectrometry : RCM, 32(21), 1831-1842 (2018-06-27)
Carry-over is an undesirable contamination from medicated to non-medicated during the production of feedingstuffs. In 2014 the European Parliament and the Council started working to produce a new regulatory act that will fix tolerable levels of drugs by carry-over in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service