Skip to Content
Merck
All Photos(1)

Key Documents

33743

Supelco

Chlozolinate solution

100 μg/mL in cyclohexane, PESTANAL®, analytical standard

Synonym(s):

Ethyl 3-(3,5-dichlorophenyl)-5- methyl-2,4-dioxo-5-oxazolidinecarboxyate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11Cl2NO5
CAS Number:
Molecular Weight:
332.14
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in cyclohexane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

InChI

1S/C13H11Cl2NO5/c1-3-20-11(18)13(2)10(17)16(12(19)21-13)9-5-7(14)4-8(15)6-9/h4-6H,3H2,1-2H3

InChI key

IGUYEXXAGBDLLX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chlozolinate is a systemic fungicide, used for the control of downy mildew, brown rot, and fruit rot in agricultural commodities.[1]

Application

Chlozolinate may be used as an analytical reference standard for the determination of the analyte in food commodities using gas and liquid chromatography coupled to tandem mass spectrometry.[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of multiclass pesticides in food commodities by pressurized liquid extraction using GC-MS/MS and LC-MS/MS.
Frenich G.A, et al.
Analytical and Bioanalytical Chemistry, 383(7-8), 1106-1118 (2005)
Part 2: Insecticides and Fungicides.
Metabolic Pathways of Agrochemicals: Herbicides and plant growth regulators;, 383(7-8), 1106-1118 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service