Skip to Content
Merck
All Photos(1)

Key Documents

33437

Sigma-Aldrich

Ninhydrin

puriss. p.a., ≥99%

Synonym(s):

1,2,3-Indantrione monohydrate, 2,2-Dihydroxy-1,3-indanedione, Trioxohydrindene monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6O4
CAS Number:
Molecular Weight:
178.14
Beilstein:
1910963
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

puriss. p.a.

Assay

≥99%

ign. residue

≤0.1% (as SO4)

pH

4.6-5.0 (20 °C, 1%)

mp

250 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

OC1(O)C(=O)c2ccccc2C1=O

InChI

1S/C9H6O4/c10-7-5-3-1-2-4-6(5)8(11)9(7,12)13/h1-4,12-13H

InChI key

FEMOMIGRRWSMCU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ninhydrin (triketohydrindene hydrate) may be used as reagent for the quantification of amino acids and peptides by photometric method and colorimetric method. It may be used to prepare the ninhydrin reagent, by mixing ninhydrin and hydrindantin (reduced form of ninhydrin) in dimethyl sulfoxide. This reagent is used in manual ninhydrin method for the quantitative determination of amino acids.
Used for the detection of free amino groups in amino acids, peptides and proteins.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photometric ninhydrin method for use in the chromatography of amino acids.
S MOORE et al.
The Journal of biological chemistry, 176(1), 367-388 (1948-10-01)
Yu-Feng Xie et al.
Journal of neurochemistry, 122(3), 605-618 (2012-05-23)
Neurons located in the trigeminal subnucleus caudalis (Vc) play crucial roles in pain and sensorimotor functions in the orofacial region. Because of many anatomical and functional similarities with the spinal dorsal horn (SDH), Vc has been termed the medullary dorsal
Joseph Almog et al.
Journal of forensic sciences, 56 Suppl 1, S162-S165 (2011-01-14)
We explored the quality distribution of ninhydrin-developed prints on A4 bond paper in two groups of individuals, in Israel and in India. While the quality distributions of the developed marks in both countries had some dissimilarities, both groups showed the
The determination of amino-acids with ninhydrin.
Yemm EW, et al.
Analyst, 80(948), 209-214 (1955)
Mendel Friedman
Journal of agricultural and food chemistry, 52(3), 385-406 (2004-02-05)
The reaction of ninhydrin with primary amino groups to form the purple dye now called Ruhemann's purple (RP) was discovered by Siegfried Ruhemann in 1910. In addition, imines such as pipecolic acid and proline, the guanidino group of arginine, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service