Skip to Content
Merck
All Photos(1)

Key Documents

33415

Supelco

Aflatoxin Mix 4 solution

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
85151701
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

multi-component solution

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Aflatoxins are a group of hypertoxic secondary metabolites produced by fungi of the genus Aspergillus, mostly A. flavus, A. Parasiticus, and A. nomius. Of the 20 aflatoxins identified so far, B1, B2, G1, and G2 are the four naturally occurring aflatoxins.

Application

It is intended to be used for the calibration of analytical instruments, validation of analytical methods, determination of detection limits and linearity studies, and recovery experiments.

This aflatoxin mix 4 solution was used as follows:
  • Ultra-high-performance liquid chromatography-fluorescence detection (UHPLC-FLD) based quantification of aflatoxins— B1, B2, G1, and G2, in the coffee bean and powder samples
  • Estimation of aflatoxins (B1, B2, G1, and G2) in samples of maize, rice, sorghum meal, and peanuts by ultra high-performance liquid chromatography coupled with a fluorescence detector
  • Determination of naturally occurring aflatoxins in 65 peanut-enriched flour samples by high-performance liquid chromatography (HPLC) combined with fluorescence detection

Components

B1, G1, B2, G2 in acetonitrile (20 ug/mL each)

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clustered pathway genes in aflatoxin biosynthesis.
Yu J, et al.
Applied and Environmental Microbiology, 70(3), 1253-1262 (2004)

Articles

Cannabis testing regulations include analysis of purity, potency, and contaminants. Learn more about analysis of cannabis for pesticides, mycotoxins, and pyrethrins registered in California by LC-MS/MS and GC-MS/MS Using Ascentis® Express Biphenyl and SLB®-5ms Columns

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service